4-Ethoxybenzaldehyde can be used as an anti-inflammatory and prostaglandin E2 suppressor. benzaldehyde-derived anti-inflammatory agents may be useful in reducing facial erythema.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Ethoxybenzaldehyde can be used as an anti-inflammatory and prostaglandin E2 suppressor. benzaldehyde-derived anti-inflammatory agents may be useful in reducing facial erythema.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents. Air sensitive.
RUO – Research Use Only
General References:
- Zoe Diana Draelos.; Bryan B. Fuller. Efficacy of 1% 4-Ethoxybenzaldehyde in Reducing Facial Erythema. Dermatologic Surgery. 2005, 31 881-886,.
- M. P. M. Marques .; A. M. Amorim da Costa.; Paulo J. A. Ribeiro-Claro. Evidence of C-H···O Hydrogen Bonds in Liquid 4-Ethoxybenzaldehyde by NMR and Vibrational Spectroscopies. J. Phys. Chem. 2001, 105 (21),5292-5297.