1x1 image pixel for data collection
N,N-Dimethyl-p-toluidine, 99%
N,N-Dimethyl-p-toluidine, 99%
N,N-Dimethyl-p-toluidine, 99%
N,N-Dimethyl-p-toluidine, 99%
Thermo Scientific Chemicals

N,N-Dimethyl-p-toluidine, 99%

CAS: 99-97-8 | C9H13N | 135.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A11988.0E
also known as A11988-0E
Price (USD)
511.65
Online Exclusive
568.00
Save 56.35 (10%)
Each
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)
511.65
Online Exclusive
568.00
Save 56.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS99-97-8
IUPAC NameN,N,4-trimethylaniline
Molecular FormulaC9H13N
InChI KeyGYVGXEWAOAAJEU-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, yellow
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.5445-1.5475 @ 20?C
FormLiquid
N,N-Dimethyl-p-toluidine is used as a polymerization catalyst for polyesters, acrylate and epoxy resins. It is also used as a hardener for dental cements and in adhesives. It serves as an intermediate for photographic chemicals, in industrial glues, in artificial fingernail preparations, colorants, pharmaceuticals. It reacts with vinyl ether in the presence of copper(II) chloride gives tetrahydroquinolines. Further, it is used to accelerate polymerization of ethyl methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-p-toluidine is used as a polymerization catalyst for polyesters, acrylate and epoxy resins. It is also used as a hardener for dental cements and in adhesives. It serves as an intermediate for photographic chemicals, in industrial glues, in artificial fingernail preparations, colorants, pharmaceuticals. It reacts with vinyl ether in the presence of copper(II) chloride gives tetrahydroquinolines. Further, it is used to accelerate polymerization of ethyl methacrylate.

Solubility
Miscible with alcohol, ether and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Sakai, N.; Sasaki, M.; Ogiwara, Y. Copper(II)-catalyzed oxidative N-nitrosation of secondary and tertiary amines with nitromethane under an oxygen atmosphere. Chem. Commun. 2015, 51 (58), 11638-11641.
  2. Balta, D. K.; Karahan, Ö.; Avci, D.; Arsu, N. Synthesis, photophysical and photochemical studies of benzophenone based novel monomeric and polymeric photoinitiators. Prog. Org. Coat. 2015, 78, 200-207.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link