Thermo Scientific Chemicals

2-Deoxy-D-ribose, 99%

Catalog number: A11990.03
1 g, Each
Thermo Scientific Chemicals

2-Deoxy-D-ribose, 99%

Catalog number: A11990.03
1 g, Each
Quantity
Catalog number: A11990.03
also known as A11990-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
533-67-5
IUPAC Name
(4S,5R)-5-(hydroxymethyl)oxolane-2,4-diol
Molecular Formula
C5H10O4
InChI Key
PDWIQYODPROSQH-GLULIZFNNA-N
SMILES
OC[C@H]1OC(O)C[C@@H]1O
Appearance (Color)
White to cream or pale yellow
Form
Powder
Assay from Supplier's CofA
≥98.5 area-% (HPLC)
Identification (FTIR)
Conforms
Optical Rotation
-59.0° to -54.0° (c=1 in water, 24hr)

Description

2-Deoxy-D-ribose is used as a precursor to deoxyribonucleic acid. It induces apoptosis by inhibiting the synthesis and increasing the efflux of glutathione. Further, it is used in the synthesis of optically active dipyrrolyl alkanols from pyrroles on the surface of montmorillonite KSF clay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Deoxy-D-ribose is used as a precursor to deoxyribonucleic acid. It induces apoptosis by inhibiting the synthesis and increasing the efflux of glutathione. Further, it is used in the synthesis of optically active dipyrrolyl alkanols from pyrroles on the surface of montmorillonite KSF clay.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text