Thermo Scientific Chemicals

Diethylaminosulfur trifluoride, 95%, Thermo Scientific Chemicals

Catalog number: A11992.06
5 g, Each
Thermo Scientific Chemicals

Diethylaminosulfur trifluoride, 95%, Thermo Scientific Chemicals

Catalog number: A11992.06
5 g, Each
Quantity
Catalog number: A11992.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38078-09-0
IUPAC Name
diethyl(trifluoro-λ⁴-sulfanyl)amine
Molecular Formula
C4H10F3NS
InChI Key
CSJLBAMHHLJAAS-UHFFFAOYSA-N
SMILES
CCN(CC)S(F)(F)F
Appearance (Color)
Clear colorless to yellow to pale orange
Form
Liquid
Assay (Non-aqueous acid-base Titration)
≥94.0 to ≤106.0%
Refractive Index
1.4085-1.4145 @ 20?C

Description

Diethylaminosulfur trifluoride (DAST) is used as a precursor to prepare 2,6-dimethoxybenzoyl fluoride and 2-thiazolines. It is a useful fluorinating agent used for a variety of compounds including alcohols, thioethers, alkenols and cyanohydrins. It also serves as a catalyst for Friedel-Crafts allylation reaction using tertiary cyclopropyl silyl ethers and in the rearrangement of homoallylic alcohols to unsaturated aldehydes. It plays an important role for gem difluorination of ketopipecolinic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylaminosulfur trifluoride (DAST) is used as a precursor to prepare 2,6-dimethoxybenzoyl fluoride and 2-thiazolines. It is a useful fluorinating agent used for a variety of compounds including alcohols, thioethers, alkenols and cyanohydrins. It also serves as a catalyst for Friedel-Crafts allylation reaction using tertiary cyclopropyl silyl ethers and in the rearrangement of homoallylic alcohols to unsaturated aldehydes. It plays an important role for gem difluorination of ketopipecolinic acids.

Solubility
Miscible with water, ethanol, benzene and acetonitrile.

Notes
Store in cool place. Moisture sensitive. It reacts with water and ethanol. Incompatible with water, acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text