L-Leucine-4-nitroanilide, 99%, Thermo Scientific Chemicals
L-Leucine-4-nitroanilide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-Leucine-4-nitroanilide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5g
1g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11994.06
also known as A11994-06
Price (USD)/ Each
166.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
196.00 
Save 29.35 (15%)
-
Add to cart
Quantity:
5g
Request bulk or custom format
Price (USD)/ Each
166.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
196.00 
Save 29.35 (15%)
Add to cart
L-Leucine-4-nitroanilide, 99%, Thermo Scientific Chemicals
Catalog numberA11994.06
Price (USD)/ Each
166.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
196.00 
Save 29.35 (15%)
-
Add to cart
Chemical Identifiers
CAS4178-93-2
IUPAC Name(2S)-2-amino-4-methyl-N-(4-nitrophenyl)pentanamide
Molecular FormulaC12H17N3O3
InChI KeyAXZJHDNQDSVIDR-NSHDSACASA-N
SMILESCC(C)C[C@H](N)C(=O)NC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5% (non-U.S. specification)
Appearance (Color)White to cream to pale yellow
Assay from Suppliers CofA≥98.5% (HPLC, U.S. specification)
Identification (FTIR)Conforms (non-U.S. specification)
View more
It is used to detect/evaluate functionality of amino peptidases. The compound is used in an ELISA assay to detect biological functionality of chicken amino peptidase produced by E. coli. It is also employed to characterize the native IRAP (placental leucine amino peptidase). Leu-pNA, Chromogenic Substrate is documented for use with pure samples as well as cell extracts in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to detect/evaluate functionality of amino peptidases. The compound is used in an ELISA assay to detect biological functionality of chicken amino peptidase produced by E. coli. It is also employed to characterize the native IRAP (placental leucine amino peptidase). Leu-pNA, Chromogenic Substrate is documented for use with pure samples as well as cell extracts in vitro.

Solubility
Soluble in DMSO (>100 mM), and methanol (>25 mM).

Notes
Store away from oxidizing agents. Keep the container tightly closed in a well ventilated place.
RUO – Research Use Only
A Bernkop-Schnürch.; M K Marschütz. Development and in vitro evaluation of systems to protect peptide drugs from aminopeptidase N.Pharm. Res.1997,14(2), 181-185.

E K Deniakina.; A D Nekliudov.; T A Loginova.; I A Krest’ianova.; Iu E Bartoshevich. Production and properties of an immobilized enzyme preparation with peptidase activity.Prikl Biokhim Mikrobiol.1985,21(2), 177-83.