Thermo Scientific Chemicals

Tetra-n-propylammonium hydrogen sulfate, 98%, Thermo Scientific Chemicals

Catalog number: A12002.06
5 g, Each
Thermo Scientific Chemicals

Tetra-n-propylammonium hydrogen sulfate, 98%, Thermo Scientific Chemicals

Catalog number: A12002.06
5 g, Each
Quantity
Catalog number: A12002.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56211-70-2
IUPAC Name
tetrapropylazanium hydrogen sulfate
Molecular Formula
C12H29NO4S
InChI Key
MOXJKKOSZCHGEU-UHFFFAOYSA-M
SMILES
OS([O-])(=O)=O.CCC[N+](CCC)(CCC)CCC
Appearance (Color)
White
Form
Powder or crystalline powder and/or lumps
Assay (Aqueous acid-base Titration)
≥97.5 to <102.5%
Water Content (Karl Fischer Titration)
<1.0%
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text