Thermo Scientific Chemicals

Hexa-n-butylditin, 97%, Thermo Scientific Chemicals

Catalog number: A12007.09
10 g, Each
Thermo Scientific Chemicals

Hexa-n-butylditin, 97%, Thermo Scientific Chemicals

Catalog number: A12007.09
10 g, Each
Quantity
Catalog number: A12007.09
also known as A12007-09
Price (USD)
Have Questions?

Chemical Identifiers

CAS
813-19-4
IUPAC Name
hexabutyldistannane
Molecular Formula
C24H54Sn2
InChI Key
REDSKZBUUUQMSK-UHFFFAOYSA-N
SMILES
CCCC[Sn](CCCC)(CCCC)[Sn](CCCC)(CCCC)CCCC
Assay (GC)
≥96.0%
Refractive Index
1.5105-1.5145 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid

Description

Hexa-n-butylditin is used in palladium-catalyzed tin-carbon bond formation. It finds useful application in deoxygenation and desulfurization reactions. It acts as a source of tributylstannyl radicals in synthetic chemistry. It is also used in the radioiodination of styrlpyridines. Further, it is used as a reagent to stannylate aryl halides for further Stille coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexa-n-butylditin is used in palladium-catalyzed tin-carbon bond formation. It finds useful application in deoxygenation and desulfurization reactions. It acts as a source of tributylstannyl radicals in synthetic chemistry. It is also used in the radioiodination of styrlpyridines. Further, it is used as a reagent to stannylate aryl halides for further Stille coupling.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text