Thermo Scientific Chemicals

4-Methyl-2-nitrophenol, 97%, Thermo Scientific Chemicals

Catalog number: A12009.14
25 g, Each
Thermo Scientific Chemicals

4-Methyl-2-nitrophenol, 97%, Thermo Scientific Chemicals

Catalog number: A12009.14
25 g, Each
Quantity
Catalog number: A12009.14
also known as A12009-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
119-33-5
IUPAC Name
4-methyl-2-nitrophenol
Molecular Formula
C7H7NO3
InChI Key
SYDNSSSQVSOXTN-UHFFFAOYSA-N
SMILES
CC1=CC=C(O)C(=C1)[N+]([O-])=O
Appearance (Color)
Dark yellow to yellow to brown
Form
Fused solid or clear liquid as melt or chunks
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
27.0-36.0°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text