The derivative of this compound, trans-Cinnamoylimidazole is involved in the synthesis of trans-Cinnamoyl-Papain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The derivative of this compound, trans-Cinnamoylimidazole is involved in the synthesis of trans-Cinnamoyl-Papain.
Solubility
Reacts with water.
Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases. Moisture sensitive.
RUO – Research Use Only
Lewis J. Brubacher.; Myron L. Bender. The Preparation and Properties of trans-Cinnamoyl-Papain. J. Am. Chem. Soc. 1966, 88 (24),5871-5880.
Stefan Latza.; Dietmar Gansser.; Ralf G. Berger. Identification and Accumulation of 1-O-trans-Cinnamoyl-β-d-glucopyranose in Developing Strawberry Fruit (Fragaria ananassa Duch. Cv. Kent). J. Agric. Food Chem. 1996, 44 (6),1367-1370.