Thermo Scientific Chemicals

N-(Chloromethyl)phthalimide, 97%, Thermo Scientific Chemicals

Catalog number: A12018.22
100 g, Each
Thermo Scientific Chemicals

N-(Chloromethyl)phthalimide, 97%, Thermo Scientific Chemicals

Catalog number: A12018.22
100 g, Each
Quantity
Catalog number: A12018.22
also known as A12018-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17564-64-6
IUPAC Name
2-(chloromethyl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular Formula
C9H6ClNO2
InChI Key
JKGLRGGCGUQNEX-UHFFFAOYSA-N
SMILES
ClCN1C(=O)C2=CC=CC=C2C1=O
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Assay (GC)
≥96.0%
Melting Point (clear melt)
129.0-139.0?C

Description

It is used as an agrochemical and medicine intermediates and is used in organic synthesis. Microporous membranes prepared from chemically modified polysulfone, which is a derivative of this compound have better streaming potential.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an agrochemical and medicine intermediates and is used in organic synthesis. Microporous membranes prepared from chemically modified polysulfone, which is a derivative of this compound have better streaming potential.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text