Thermo Scientific Chemicals

1-Octadecanol, 97%, Thermo Scientific Chemicals

Catalog number: A12020.0E
2500 g, Each
Thermo Scientific Chemicals

1-Octadecanol, 97%, Thermo Scientific Chemicals

Catalog number: A12020.0E
2500 g, Each
Quantity
Catalog number: A12020.0E
also known as A12020-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-92-5
IUPAC Name
octadecan-1-ol
Molecular Formula
C18H38O
InChI Key
GLDOVTGHNKAZLK-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCO
Appearance (Color)
White
Form
Flakes or pellets
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
54.0-63.0?C

Description

1-Octadecanol has been used as surfactant for the functionalization of carbon nanotubes. It is employed as emulsion stabilizer for creams and lotions, quality modifier of lipsticks, additive for ointment base and cream conditioners for smooth touch to skin for cosmetic uses. It is auxiliary for emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octadecanol has been used as surfactant for the functionalization of carbon nanotubes. It is employed as emulsion stabilizer for creams and lotions, quality modifier of lipsticks, additive for ointment base and cream conditioners for smooth touch to skin for cosmetic uses. It is auxiliary for emulsifiers.

Solubility
Insoluble in water. Soluble in alcohol, acetone, ether, benzene, chloroform.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a well ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text