Thermo Scientific Chemicals

4-Chlorothiobenzamide, 97%, Thermo Scientific Chemicals

Catalog number: A12021.22
100 g, Each
Thermo Scientific Chemicals

4-Chlorothiobenzamide, 97%, Thermo Scientific Chemicals

Catalog number: A12021.22
100 g, Each
Quantity
Catalog number: A12021.22
also known as A12021-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2521-24-6
IUPAC Name
4-chlorobenzene-1-carbothioamide
Molecular Formula
C7H6ClNS
InChI Key
OKPUICCJRDBRJT-UHFFFAOYSA-N
SMILES
NC(=S)C1=CC=C(Cl)C=C1
Form
Powder
Assay (HPLC)
≥97.5%
Appearance (Color)
Yellow to brown
Melting Point (clear melt)
121.5-130.5?C

Description

4-Chlorothiobenzamide is used as a catalytic agent and as petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorothiobenzamide is used as a catalytic agent and as petrochemical additive.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text