Thermo Scientific Chemicals

(4-Carboxybutyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: A12023.14
25 g, Each
Thermo Scientific Chemicals

(4-Carboxybutyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: A12023.14
25 g, Each
Quantity
Catalog number: A12023.14
also known as A12023-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17814-85-6
IUPAC Name
(4-carboxybutyl)triphenylphosphanium bromide
Molecular Formula
C23H24BrO2P
InChI Key
MLOSJPZSZWUDSK-UHFFFAOYSA-N
SMILES
[Br-].OC(=O)CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Solid
Appearance (Color)
White to pale cream to pale yellow
Purity
≥97.5% (HPLC)

Description

Phosphonium salt is a key intermediate for manufacture of some types of prostaglandins, which are subsequently used as active ingredients of both human and veterinary drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphonium salt is a key intermediate for manufacture of some types of prostaglandins, which are subsequently used as active ingredients of both human and veterinary drugs.

Solubility
Soluble in ethanol, methanol and soluble water. Insoluble in toluene and hexane.

Notes
Hygroscopic. Store in dry place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text