Thermo Scientific Chemicals

2-Aminothiazole, 97%, Thermo Scientific Chemicals

Catalog number: A12026.30
250 g, Each
Thermo Scientific Chemicals

2-Aminothiazole, 97%, Thermo Scientific Chemicals

Catalog number: A12026.30
250 g, Each
Quantity
Catalog number: A12026.30
also known as A12026-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-50-4
IUPAC Name
1,3-thiazol-2-amine
Molecular Formula
C3H4N2S
InChI Key
RAIPHJJURHTUIC-UHFFFAOYSA-N
SMILES
NC1=NC=CS1
Appearance (Color)
Cream to dark brown to brown
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms

Description

2-Aminothiazole is used in the synthesis of 2-aminothiazole-modified silica gel and also used in Ulmann coupling with 2-chlorobenzoic acids mediated by ultrasonic irradiation. It acts as a component of disperse dye, thyroid inhibitor in the treatment of hyperthyroidism, as the acid tartrate and as a therapeutic drug for prion diseases. It is also used to kill bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, alcohol and ether.

Notes
Incompatible with strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, nitric acid and sulfuric acid.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text