2-Benzoylthiophene, 98%, Thermo Scientific Chemicals
2-Benzoylthiophene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Benzoylthiophene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Catalog number A12036.06
also known as A12036-06
Price (USD)/ Each
38.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
38.40
Add to cart
2-Benzoylthiophene, 98%, Thermo Scientific Chemicals
Catalog numberA12036.06
Price (USD)/ Each
38.40
-
Add to cart
Chemical Identifiers
CAS135-00-2
IUPAC Namephenyl(thiophen-2-yl)methanone
Molecular FormulaC11H8OS
InChI KeyDWYFUJJWTRPARQ-UHFFFAOYSA-N
SMILESO=C(C1=CC=CS1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps or fused solid
Melting Point (clear melt)53.0-60.0?C
Assay (GC)≥97.5%
Appearance (Color)Pale cream to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only