Thermo Scientific Chemicals

Pyridoxine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A12041.22
100 g, Each
Thermo Scientific Chemicals

Pyridoxine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A12041.22
100 g, Each
Quantity
Catalog number: A12041.22
also known as A12041-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58-56-0
IUPAC Name
hydrogen 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol chloride
Molecular Formula
C8H12ClNO3
InChI Key
ZUFQODAHGAHPFQ-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CC1=NC=C(CO)C(CO)=C1O
Melting Point
200-210°C (non-U.S. sourced material)
Appearance (Color)
White to pale cream
Form
Powder or crystals or crystalline powder
Assay (Titration ex Chloride)
≥98.5 to ≤101.5% (non-U.S. sourced material)
Assay from Supplier's CofA
≥98.5% (U.S. sourced material)

Description

Pyridoxol hydrochlroride is a salt of pyridoxine free base which is used to treat low levels of vitamin B6. It is essential to maintain the health of nerves, skin and red blood cells. It is also used to treat nerve disorder and hereditary disorders like xanthurenic aciduria, cystathioninuria, hyperoxaluria and homocystinuria. It finds application in cosmetics, food and feed additives. It plays an important role in transporting protein Bsu1 and thiamine uptake. Furthermore, it is also involved in various biochemical reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridoxol hydrochlroride is a salt of pyridoxine free base which is used to treat low levels of vitamin B6. It is essential to maintain the health of nerves, skin and red blood cells. It is also used to treat nerve disorder and hereditary disorders like xanthurenic aciduria, cystathioninuria, hyperoxaluria and homocystinuria. It finds application in cosmetics, food and feed additives. It plays an important role in transporting protein Bsu1 and thiamine uptake. Furthermore, it is also involved in various biochemical reactions.

Solubility
Soluble in water and alcohol. Slightly soluble in acetone. In soluble in ether and chloroform.

Notes
Light sensitive. Incompatible with bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text