Thermo Scientific Chemicals

Biphenyl-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12049.14
25 g, Each
Thermo Scientific Chemicals

Biphenyl-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12049.14
25 g, Each
Quantity
Catalog number: A12049.14
also known as A12049-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
947-84-2
IUPAC Name
[1,1'-biphenyl]-2-carboxylic acid
Molecular Formula
C13H10O2
InChI Key
ILYSAKHOYBPSPC-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=C1C1=CC=CC=C1
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
110.0-116.0?C
Assay (Silylated GC)
≥97.5%

Description

It is used as a pharmaceutical intermediate. he reaction of 2?-substituted biphenyl-2-carboxylic acids (where the 2?-substituent is H, CO2H, NO2, Cl, OMe, or CO2Me) with lead tetra-acetate in refluxing benzene solution, under a nitrogen atmosphere, affords 3,4-benzocoumarin as a major organic product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate. he reaction of 2′-substituted biphenyl-2-carboxylic acids (where the 2′-substituent is H, CO2H, NO2, Cl, OMe, or CO2Me) with lead tetra-acetate in refluxing benzene solution, under a nitrogen atmosphere, affords 3,4-benzocoumarin as a major organic product.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text