Thermo Scientific Chemicals

2-Bromo-1,4-dichlorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A12056.14
25 g, Each
Thermo Scientific Chemicals

2-Bromo-1,4-dichlorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A12056.14
25 g, Each
Quantity
Catalog number: A12056.14
also known as A12056-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1435-50-3
IUPAC Name
2-bromo-1,4-dichlorobenzene
Molecular Formula
C6H3BrCl2
InChI Key
OVXVQBCRONSPDC-UHFFFAOYSA-N
SMILES
ClC1=CC=C(Cl)C(Br)=C1
Assay (GC)
≥97.5%
Appearance (Color)
Colorless to cream to pale brown
Melting Point (clear melt)
30.0-37.0?C
Form
Crystals or powder or crystalline powder or clear liquid as melt

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text