Thermo Scientific Chemicals

Acetylacetaldehyde dimethyl acetal, tech. 90%, Thermo Scientific Chemicals

Catalog number: A12058.0B
1000 g, Each
Thermo Scientific Chemicals

Acetylacetaldehyde dimethyl acetal, tech. 90%, Thermo Scientific Chemicals

Catalog number: A12058.0B
1000 g, Each
Quantity
Catalog number: A12058.0B
also known as A12058-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5436-21-5
IUPAC Name
4,4-dimethoxybutan-2-one
Molecular Formula
C6H12O3
InChI Key
PJCCSZUMZMCWSX-UHFFFAOYSA-N
SMILES
COC(CC(C)=O)OC
Appearance (Color)
Clear, colourless to yellow
Assay (GC)
> 88.0%
Form
liquid
Refractive Index
1.4110 - 1.4190 @20?C

Description

Acetaldehyde dimethyl acetal is used as a flavoring agent. It is also used in the preparation of (R)-4,4-dimethoxy-2-butanol, pyrazoles and pyrimidines. Further, it is used in the preparation of N,N-diethyl-[2-(4-fluorophenyl)-5-methylpyrazolo[1,5-a]-pyrimidin-3-yl]acetamide and acetoacetaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetaldehyde dimethyl acetal is used as a flavoring agent. It is also used in the preparation of (R)-4,4-dimethoxy-2-butanol, pyrazoles and pyrimidines. Further, it is used in the preparation of N,N-diethyl-[2-(4-fluorophenyl)-5-methylpyrazolo[1,5-a]-pyrimidin-3-yl]acetamide and acetoacetaldehyde.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with acids and strong oxidizing agents
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text