Thermo Scientific Chemicals

3-(4-Hydroxy-3-methoxyphenyl)propionic acid, 97%, Thermo Scientific Chemicals

Catalog number: A12069.03
1 g, Each
Thermo Scientific Chemicals

3-(4-Hydroxy-3-methoxyphenyl)propionic acid, 97%, Thermo Scientific Chemicals

Catalog number: A12069.03
1 g, Each
Quantity
Catalog number: A12069.03
also known as A12069-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1135-23-5
IUPAC Name
3-(4-hydroxy-3-methoxyphenyl)propanoic acid
Molecular Formula
C10H12O4
InChI Key
BOLQJTPHPSDZHR-UHFFFAOYSA-N
SMILES
COC1=CC(CCC(O)=O)=CC=C1O
Appearance (Color)
White to yellow to orange to brown
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
85.0-95.0°C
Form
Powder

Description

It is a caffeine metabolite which showed high antioxidant activity. It is a very sensitive biomarker for the consumption of relatively small amount of coffee. an be used to inhibit prostaglandin E(2) production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a caffeine metabolite which showed high antioxidant activity. It is a very sensitive biomarker for the consumption of relatively small amount of coffee. an be used to inhibit prostaglandin E(2) production.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Store in cool place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text