Thermo Scientific Chemicals

Cacodylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12075.88
2.5 g, Each
Thermo Scientific Chemicals

Cacodylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A12075.88
2.5 g, Each
Quantity
Catalog number: A12075.88
also known as A12075-88
Price (USD)
Have Questions?

Chemical Identifiers

CAS
75-60-5
IUPAC Name
dimethylarsinic acid
Molecular Formula
C2H7AsO2
InChI Key
OGGXGZAMXPVRFZ-UHFFFAOYSA-N
SMILES
C[As](C)(O)=O
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
195.0-201.0?C
Identification (FTIR)
Conforms

Description

A useful arsenic acid for proteomics research. It a good substitute for phosphate in applications to avoid phosphates. Cacodylic Acid is useful for some DNA applications and is also popular in microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful arsenic acid for proteomics research. It a good substitute for phosphate in applications to avoid phosphates. Cacodylic Acid is useful for some DNA applications and is also popular in microscopy.

Solubility
Soluble in water (50 mg/ml), water (2000 at 25°C), alcohols, and acetic acid. Insoluble in ether.

Notes
Hygroscopic. Store away from moisture, air and oxidizing agents. Store under inert gas.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text