Appearance (Color)Clear colorless to very pale yellow
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.4115-1.4145 @ 20°C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.