Thermo Scientific Chemicals

1-Octen-3-ol, 98%, Thermo Scientific Chemicals

Catalog number: A12079.30
250 g, Each
Thermo Scientific Chemicals

1-Octen-3-ol, 98%, Thermo Scientific Chemicals

Catalog number: A12079.30
250 g, Each
Quantity
Catalog number: A12079.30
also known as A12079-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3391-86-4
IUPAC Name
oct-1-en-3-ol
Molecular Formula
C8H16O
InChI Key
VSMOENVRRABVKN-UHFFFAOYNA-N
SMILES
CCCCCC(O)C=C
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Refractive Index
1.4355-1.4395 @ 20?C
Form
Liquid

Description

Used as a flavoring agent in cheese and chocolate. Used as a perfuming agent in cosmetic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a flavoring agent in cheese and chocolate. Used as a perfuming agent in cosmetic industry.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text