Triallylamine, 97%, Thermo Scientific Chemicals
Triallylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triallylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
250 g
Catalog number A12081.30
also known as A12081-30
Price (USD)/ Each
112.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
112.00
Add to cart
Triallylamine, 97%, Thermo Scientific Chemicals
Catalog numberA12081.30
Price (USD)/ Each
112.00
-
Add to cart
Chemical Identifiers
CAS102-70-5
IUPAC Nametris(prop-2-en-1-yl)amine
Molecular FormulaC9H15N
InChI KeyVPYJNCGUESNPMV-UHFFFAOYSA-N
SMILESC=CCN(CC=C)CC=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥96.0%
Refractive Index1.4500-1.4550 @ 20?C
It is employed in the production of other chemicals. It is also mainly applied in organic synthesis and resin modifiers. It is used in the cross linking of high absorbent and the intermediates of ion exchange resin. It can also be employed in producing polyester activator and the evocating agent of butadiene polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the production of other chemicals. It is also mainly applied in organic synthesis and resin modifiers. It is used in the cross linking of high absorbent and the intermediates of ion exchange resin. It can also be employed in producing polyester activator and the evocating agent of butadiene polymerization.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from strong oxidizing agents and acids. Protect against electrostatic charges and keep away from ignition sources.
RUO – Research Use Only

General References:

  1. H G Swarts.; C H Klaassen.; F M Schuurmans Stekhoven.; J J De Pont. Tertiary amines as antagonists of both the luminal and cytosolic K(+)-site of gastric H,K-ATPase.Biochim Biophys Acta.1994,1189(2), 143-151.
  2. E D Thompson.; J L Seymour.; M J Aardema.; R A LeBoeuf.; B L Evans.; D B Cody. Lack of genotoxicity of cross-linked acrylate polymers in four short-term genotoxicity assays.Environ Mol Mutagen.1991,18(3), 184-199.