Succinic acid, 99+%, Thermo Scientific Chemicals
Succinic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Succinic acid, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
500 g
10,000 g
Catalog number A12084.0E
also known as A12084-0E
Price (USD)/ Each
168.65
Online exclusive
187.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
168.65
Online exclusive
187.00 
Save 18.35 (10%)
Add to cart
Succinic acid, 99+%, Thermo Scientific Chemicals
Catalog numberA12084.0E
Price (USD)/ Each
168.65
Online exclusive
187.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS110-15-6
IUPAC Namebutanedioic acid
Molecular FormulaC4H6O4
InChI KeyKDYFGRWQOYBRFD-UHFFFAOYSA-N
SMILESOC(=O)CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Identification (FTIR)Conforms
Assay (Aqueous acid-base Titration)≥99.0 to ≤101.0%
FormCrystals or powder or crystalline powder
Succinic acid acts as a leaching agent in extracting lithium, cobalt from used Li-ion batteries and magnesium from magnesite ore. It is used as a starting material in the synthesis of new elastic polyesters, fumaric acid, succinic anhydride and alkyd resins. It plays an important role as an acidity regulator in the food and beverage industry, as an excipient in pharmaceutical products and acts as a cocrystallising agent in organic synthesis. Its derivative viz diethyl ester is used as a substrate in the Stobbe condensation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Succinic acid acts as a leaching agent in extracting lithium, cobalt from used Li-ion batteries and magnesium from magnesite ore. It is used as a starting material in the synthesis of new elastic polyesters, fumaric acid, succinic anhydride and alkyd resins. It plays an important role as an acidity regulator in the food and beverage industry, as an excipient in pharmaceutical products and acts as a cocrystallising agent in organic synthesis. Its derivative viz diethyl ester is used as a substrate in the Stobbe condensation.

Solubility
Soluble in ethanol, ethyl ether, acetone and methanol. Insoluble in toluene, benzene, carbon disulfide, carbon tetrachloride and petroleum ether.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

General References:

  1. Hwang, H. J.; Choi, S. P.; Lee, S. Y.; Choi, J. I.; Han, S. J.; Lee, P. C. Dynamics of membrane fatty acid composition of succinic acid-producing Anaerobiospirillum succiniciproducens. J. Biotechnol. 2015, 193, 130-133.
  2. Gunnarsson, I. B.; Kuglarz, M.; Karakashev, D.; Angelidaki, I. Thermochemical pretreatments for enhancing succinic acid production from industrial hemp (Cannabis sativa L.). Bioresour. Technol. 2015, 182, 58-66.