Perfluoroheptanoic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Perfluoroheptanoic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only
Jing Chen.; Peng-yi Zhang.; Jian Liu. Photodegradation of perfluorooctanoic acid by 185 nm vacuum ultraviolet light. Journal of Environmental Sciences. 2007, 19 (4),387-390.
Kohtaro Ohmori.; Naomi Kudo.; Kazunori Katayama.; Yoichi Kawashim. Comparison of the toxicokinetics between perfluorocarboxylic acids with different carbon chain length. Toxicology. 2003, 184 (2-3),135-140.