Thermo Scientific Chemicals

Tri(o-tolyl)phosphine, 98+%, Thermo Scientific Chemicals

Catalog number: A12093.06
5 g, Each
Thermo Scientific Chemicals

Tri(o-tolyl)phosphine, 98+%, Thermo Scientific Chemicals

Catalog number: A12093.06
5 g, Each
Quantity
Catalog number: A12093.06
also known as A12093-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6163-58-2
IUPAC Name
tris(2-methylphenyl)phosphane
Molecular Formula
C21H21P
InChI Key
COIOYMYWGDAQPM-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1P(C1=CC=CC=C1C)C1=CC=CC=C1C
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (GC)
≥98.0%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
122.0-129.0?C

Description

Tri(o-tolyl)phosphine is used in a ruthenium-catalyzed direct amination of alcohols. It is also used in Suzuki reaction. Further, it is used in the preparation of tri-ortho-phosphinselenide by reacting with selenium as a reagent. In addition to this, it acts as a ligand in coordination chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri(o-tolyl)phosphine is used in a ruthenium-catalyzed direct amination of alcohols. It is also used in Suzuki reaction. Further, it is used in the preparation of tri-ortho-phosphinselenide by reacting with selenium as a reagent. In addition to this, it acts as a ligand in coordination chemistry.

Solubility
Soluble in alcohol. Slightly soluble in cold water. Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text