Thermo Scientific Chemicals

4-Bromobutyryl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A12094.06
5 g, Each
Thermo Scientific Chemicals

4-Bromobutyryl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A12094.06
5 g, Each
Quantity
Catalog number: A12094.06
also known as A12094-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
927-58-2
IUPAC Name
4-bromobutanoyl chloride
Molecular Formula
C4H6BrClO
InChI Key
LRTRXDSAJLSRTG-UHFFFAOYSA-N
SMILES
ClC(=O)CCCBr
Form
Liquid
Assay (GC)
≥96.0%
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4900-1.4950 @ 20?C

Description

4-Bromobutyryl chloride is used as a precursor for the synthesis of carolic acid by reacting with ethoxymagnesiomalonic ester. It is widely utilized as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobutyryl chloride is used as a precursor for the synthesis of carolic acid by reacting with ethoxymagnesiomalonic ester. It is widely utilized as an intermediate in organic synthesis and pharmaceuticals.

Solubility
Miscible with organic solvents. Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents. Remove all sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text