4-Acetoxy-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
4-Acetoxy-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Acetoxy-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
Catalog number A12103.14
also known as A12103-14
Price (USD)/ Each
47.90
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
47.90
Add to cart
4-Acetoxy-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Catalog numberA12103.14
Price (USD)/ Each
47.90
-
Add to cart
Chemical Identifiers
CAS881-68-5
IUPAC Name4-formyl-2-methoxyphenyl acetate
Molecular FormulaC10H10O4
InChI KeyPZSJOBKRSVRODF-UHFFFAOYSA-N
SMILESCOC1=CC(C=O)=CC=C1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
CommentSpecification differs for U.S. and non-U.S. material where indicated
Free acid (titration)≤1.0% (non-U.S. specification)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only