Thermo Scientific Chemicals

Isobutyraldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A12106.0F
2500 mL, Each
Thermo Scientific Chemicals

Isobutyraldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A12106.0F
2500 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A12106.0F
also known as A12106-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78-84-2
IUPAC Name
2-methylpropanal
Molecular Formula
C4H8O
InChI Key
AMIMRNSIRUDHCM-UHFFFAOYSA-N
SMILES
CC(C)C=O
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Free acid (titration)
≤2%
Identification (FTIR)
Conforms

Description

Isobutyraldehyde is used as an intermediate in the preparation of isobutanol, methacrolein, hydroxypivaldehyde and neopentyl glycol. It is actively involved in the Cannizaro reaction. It is also used as an intermediate to prepare pharmaceuticals, agrochemicals, vitamins, antioxidants, rubber accelerators, textile auxiliaries, perfumery and flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyraldehyde is used as an intermediate in the preparation of isobutanol, methacrolein, hydroxypivaldehyde and neopentyl glycol. It is actively involved in the Cannizaro reaction. It is also used as an intermediate to prepare pharmaceuticals, agrochemicals, vitamins, antioxidants, rubber accelerators, textile auxiliaries, perfumery and flavors.

Solubility
Slightly miscible with water.

Notes
Air sensitive. Store in a cool place. Incompatible with oxidizing agents, strong acids, strong bases and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text