Thermo Scientific Chemicals

3-Octanone, 98%, Thermo Scientific Chemicals

Catalog number: A12113.22
100 g, Each
Thermo Scientific Chemicals

3-Octanone, 98%, Thermo Scientific Chemicals

Catalog number: A12113.22
100 g, Each
Quantity
Catalog number: A12113.22
also known as A12113-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-68-3
IUPAC Name
octan-3-one
Molecular Formula
C8H16O
InChI Key
RHLVCLIPMVJYKS-UHFFFAOYSA-N
SMILES
CCCCCC(=O)CC
Form
Liquid
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Refractive Index
1.4140-1.4180 @ 20?C

Description

3-Octanone was used to study the effect of TiO2 photo catalyst on the rate and the ratio of products generated in photo catalytic oxidation of 3-octanone. It is used as a flavor and fragrance ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Octanone was used to study the effect of TiO2 photo catalyst on the rate and the ratio of products generated in photo catalytic oxidation of 3-octanone. It is used as a flavor and fragrance ingredient.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Incompatible with strong oxidizing agents, strong reducing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text