Thermo Scientific Chemicals

4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals

Catalog number: A12115.0B
1000 g, Each
Thermo Scientific Chemicals

4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals

Catalog number: A12115.0B
1000 g, Each
Quantity
Catalog number: A12115.0B
also known as A12115-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-91-2
IUPAC Name
1-(4-chlorophenyl)ethan-1-one
Molecular Formula
C8H7ClO
InChI Key
BUZYGTVTZYSBCU-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=C(Cl)C=C1
Form
Liquid
Refractive Index
1.5535-1.5575 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms

Description

Chloroacetophenone is used as a tear gas for riot control and as a kind of chemical mace against small groups or individuals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetophenone is used as a tear gas for riot control and as a kind of chemical mace against small groups or individuals

Solubility
Soluble in water (0.11 mg/ml at 25°C), alcohol, and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text