Thermo Scientific Chemicals

Phosphonoacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12117.09
10 g, Each
Thermo Scientific Chemicals

Phosphonoacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12117.09
10 g, Each
Quantity
Catalog number: A12117.09
also known as A12117-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4408-78-0
IUPAC Name
2-phosphonoacetic acid
Molecular Formula
C2H5O5P
InChI Key
XUYJLQHKOGNDPB-UHFFFAOYSA-N
SMILES
OC(=O)CP(O)(O)=O
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Appearance (Color)
White

Description

Phosphonoacetic acid inhibits deoxyribo nucleic acid polymerase, especially in viruses. It is used as an antiviral agent. Further, it is used in the preparation of 1-(dimethylamino)-2,2-bis[(dimethylimmonio)-methyl]ethene diperchlorate by reacting with phosphorus oxychloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphonoacetic acid inhibits deoxyribo nucleic acid polymerase, especially in viruses. It is used as an antiviral agent. Further, it is used in the preparation of 1-(dimethylamino)-2,2-bis[(dimethylimmonio)-methyl]ethene diperchlorate by reacting with phosphorus oxychloride.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text