Phosphonoacetic acid inhibits deoxyribo nucleic acid polymerase, especially in viruses. It is used as an antiviral agent. Further, it is used in the preparation of 1-(dimethylamino)-2,2-bis[(dimethylimmonio)-methyl]ethene diperchlorate by reacting with phosphorus oxychloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phosphonoacetic acid inhibits deoxyribo nucleic acid polymerase, especially in viruses. It is used as an antiviral agent. Further, it is used in the preparation of 1-(dimethylamino)-2,2-bis[(dimethylimmonio)-methyl]ethene diperchlorate by reacting with phosphorus oxychloride.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
Takenaga, A.; Kikuchi, T.; Natsui, S.; Suzuki, R. O. Self-Ordered Aluminum Anodizing in Phosphonoacetic Acid and Its Structural Coloration. ECS Solid State Lett. 2015, 4 (8), P55-P58.
Klimek-Ochab, M. Phosphate-independent utilization of phosphonoacetic acid as sole phosphorus source by a psychrophilic strain of Geomyces pannorum P15. Folia microbial. 2014, 59 (5), 375-380.