Thermo Scientific Chemicals

Benzenesulfinic acid sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: A12118.14
25 g, Each
Thermo Scientific Chemicals

Benzenesulfinic acid sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: A12118.14
25 g, Each
Quantity
Catalog number: A12118.14
also known as A12118-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
873-55-2
IUPAC Name
benzenesulfinate
Molecular Formula
C6H5O2S
InChI Key
JEHKKBHWRAXMCH-UHFFFAOYSA-M
SMILES
[O-]S(=O)C1=CC=CC=C1
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Granular solid or powder or crystalline powder or crystals
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
<0.5%

Description

It is used as a polymer adhesive enhancers, plasticizers for plasticized and modified of polyamide, epoxy resin, phenolic resin. It is designed for manufacturing of photography, pharmaceutical, electroplating industry, can be used as photo-reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a polymer adhesive enhancers, plasticizers for plasticized and modified of polyamide, epoxy resin, phenolic resin. It is designed for manufacturing of photography, pharmaceutical, electroplating industry, can be used as photo-reducing agent.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidants. Store in cool, dry place in a tightly sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text