2,3,4,5,6-Pentafluorobenzyl alcohol, 98%, Thermo Scientific Chemicals
2,3,4,5,6-Pentafluorobenzyl alcohol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3,4,5,6-Pentafluorobenzyl alcohol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A12119.14
also known as A12119-14
Price (USD)/ Each
171.65
Online exclusive
190.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
171.65
Online exclusive
190.00 
Save 18.35 (10%)
Add to cart
2,3,4,5,6-Pentafluorobenzyl alcohol, 98%, Thermo Scientific Chemicals
Catalog numberA12119.14
Price (USD)/ Each
171.65
Online exclusive
190.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS440-60-8
IUPAC Name(2,3,4,5,6-pentafluorophenyl)methanol
Molecular FormulaC7H3F5O
InChI KeyPGJYYCIOYBZTPU-UHFFFAOYSA-N
SMILESOCC1=C(F)C(F)=C(F)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or crystalline powder or fused solid
Assay (GC)≥97.5%
Melting Point (clear melt)31.5-37.5°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only