Thermo Scientific Chemicals

2,6-Dinitroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A12120.06
5 g, Each
Thermo Scientific Chemicals

2,6-Dinitroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A12120.06
5 g, Each
Quantity
Catalog number: A12120.06
also known as A12120-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
606-22-4
IUPAC Name
2,6-dinitroaniline
Molecular Formula
C6H5N3O4
InChI Key
QFUSCYRJMXLNRB-UHFFFAOYSA-N
SMILES
NC1=C(C=CC=C1[N+]([O-])=O)[N+]([O-])=O
Form
Crystals or powder or crystalline powder or flakes
Appearance (Color)
Cream to yellow to yellow-brown to brown
Assay (GC)
≥97.5%
Melting Point (clear melt)
136.0-142.0?C

Description

2,6-Dinitroaniline is used for organic synthesis intermediates, dyes intermediates, used in the preparation of 2,6-dinitroiodobenzene. It act as a precursor for various new 1,2,3-trisubstituted halobenzene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dinitroaniline is used for organic synthesis intermediates, dyes intermediates, used in the preparation of 2,6-dinitroiodobenzene. It act as a precursor for various new 1,2,3-trisubstituted halobenzene derivatives.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from strong acids, acid chlorides, acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text