4-Nitrocatechol, 98+%, Thermo Scientific Chemicals
4-Nitrocatechol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitrocatechol, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12122.06
also known as A12122-06
Price (USD)/ Each
54.65
Online exclusive
60.90 
Save 6.25 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
54.65
Online exclusive
60.90 
Save 6.25 (10%)
Add to cart
4-Nitrocatechol, 98+%, Thermo Scientific Chemicals
Catalog numberA12122.06
Price (USD)/ Each
54.65
Online exclusive
60.90 
Save 6.25 (10%)
-
Add to cart
Chemical Identifiers
CAS3316-09-4
IUPAC Name4-nitrobenzene-1,2-diol
Molecular FormulaC6H5NO4
InChI KeyXJNPNXSISMKQEX-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)172.0-178.0?C
Appearance (Color)Pale yellow to yellow to brown
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (HPLC)≥98.0%
FormCrystals or powder or crystalline powder
4-nitrocatechol (4ncat) is a suitable ligand for amperometric determination of Al(III) at environmental concentrations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-nitrocatechol (4ncat) is a suitable ligand for amperometric determination of Al(III) at environmental concentrations.

Solubility
Soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. H. Baum, K.S. Dodgson.; B. Spencer. The assay of arylsulphatases A and B in human urine. Clin. Chim. Acta. 1959, 4 (3),453-455.
  2. Ashvini Chauhan.; Asit K. Chakraborti.; Rakesh K. Jaina. Plasmid-Encoded Degradation of p-Nitrophenol and 4-Nitrocatechol by Arthrobacter protophormiae. Biochem. Biophys. Res. Commun. 2000, 270 (3),733-740.
  3. Reagent for Ge.