Calcium dihydrogen phosphate hydrate, 97%, Thermo Scientific Chemicals
Calcium dihydrogen phosphate hydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium dihydrogen phosphate hydrate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 g
500 g
2500 g
Catalog number A12123.0C
also known as A12123-0C
Price (USD)/ Each
487.65
Online exclusive
542.00 
Save 54.35 (10%)
-
Add to cart
Quantity:
10,000 g
Request bulk or custom format
Price (USD)/ Each
487.65
Online exclusive
542.00 
Save 54.35 (10%)
Add to cart
Calcium dihydrogen phosphate hydrate, 97%, Thermo Scientific Chemicals
Catalog numberA12123.0C
Price (USD)/ Each
487.65
Online exclusive
542.00 
Save 54.35 (10%)
-
Add to cart
Chemical Identifiers
CAS301524-28-7
IUPAC Namecalcium didihydrogen phosphate
Molecular FormulaCaH4O8P2
InChI KeyYYRMJZQKEFZXMX-UHFFFAOYSA-L
SMILES[Ca++].OP(O)([O-])=O.OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Elemental analysis by ICPCalcium (Ca): 16.46-17.79%
Appearance (Color)White
Assay (unspecified)(Based on Ca content): >96.0 to <104.0% (dry wt. basis)
Loss on Drying≤10% (105?C/vacuum/4hrs)
Calcium dihydrogen phosphate hydrate is a pH control reagent, stabilizer for plastics, manufacture of glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium dihydrogen phosphate hydrate is a pH control reagent, stabilizer for plastics, manufacture of glass.

Solubility
Soluble in water 18 g/L @ 30°C.

Notes
Loses H{2}O at 100°, dec. at 200°
RUO – Research Use Only

General References:

  1. Jørgen Christoffersen; Margaret R. Christoffersen; Wiktor Kibalczyc; Flemming A. Andersen. A contribution to the understanding of the formation of calcium phosphates. Journal of Crystal Growth. 1989, 94 (3), 767-777.
  2. M.S. Tung; L.C. Chow; W.E. Brown. Basic Biological Sciences Hydrolysis of Dicalcium Phosphate Dihydrate in the Presence or Absence of Calcium Fluoride. JDR. 1985, 64 (1), 2-5.