Thermo Scientific Chemicals

Diphenylphosphonic azide, 97%, Thermo Scientific Chemicals

Catalog number: A12124.06
5 g, Each
Thermo Scientific Chemicals

Diphenylphosphonic azide, 97%, Thermo Scientific Chemicals

Catalog number: A12124.06
5 g, Each
Quantity
Catalog number: A12124.06
also known as A12124-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26386-88-9
IUPAC Name
{[azido(phenoxy)phosphoryl]oxy}benzene
Molecular Formula
C12H10N3O3P
InChI Key
SORGEQQSQGNZFI-UHFFFAOYSA-N
SMILES
[N-]=[N+]=NP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥95.0%
Refractive Index
1.5485-1.5535 @ 20°C
Form
Liquid
Identification (FTIR)
Conforms

Description

Diphenylphosphonic azide acts as a reagent for the synthesis of peptides and phosphoramidates by reacting with amines. It is also used in the preparation of oligosaccharides linked with carbamate and urea bonds utilizing modified Curtis rearrangement. It is involved in pseudohalogen replacement of the azido group by treatment with nucleophilic reagents, such as water, butanol, ammonia, and various amines. Further, it is used as a hydroazidation catalyst for preparation of organoazides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylphosphonic azide acts as a reagent for the synthesis of peptides and phosphoramidates by reacting with amines. It is also used in the preparation of oligosaccharides linked with carbamate and urea bonds utilizing modified Curtis rearrangement. It is involved in pseudohalogen replacement of the azido group by treatment with nucleophilic reagents, such as water, butanol, ammonia, and various amines. Further, it is used as a hydroazidation catalyst for preparation of organoazides.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with acids and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text