2-Chlorobenzyl bromide, 97%, Thermo Scientific Chemicals
2-Chlorobenzyl bromide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chlorobenzyl bromide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A12129.14
also known as A12129-14
Price (USD)/ Each
113.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
113.00
Add to cart
2-Chlorobenzyl bromide, 97%, Thermo Scientific Chemicals
Catalog numberA12129.14
Price (USD)/ Each
113.00
-
Add to cart
Chemical Identifiers
CAS611-17-6
IUPAC Name1-(bromomethyl)-2-chlorobenzene
Molecular FormulaC7H6BrCl
InChI KeyPURSZYWBIQIANP-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to yellow
Refractive Index1.5900-1.5950 @ 20?C
Assay (GC)≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only