Thermo Scientific Chemicals

2-Chlorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: A12129.22
100 g, Each
Thermo Scientific Chemicals

2-Chlorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: A12129.22
100 g, Each
Quantity
Catalog number: A12129.22
also known as A12129-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
611-17-6
IUPAC Name
1-(bromomethyl)-2-chlorobenzene
Molecular Formula
C7H6BrCl
InChI Key
PURSZYWBIQIANP-UHFFFAOYSA-N
SMILES
ClC1=CC=CC=C1CBr
Appearance (Color)
Clear pale yellow to yellow
Assay (GC)
≥96.0%
Refractive Index
1.5900-1.5950 @ 20?C
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text