Thermo Scientific Chemicals

Ethyl 4-piperidone-3-carboxylate hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A12131.06
5 g, Each
Thermo Scientific Chemicals

Ethyl 4-piperidone-3-carboxylate hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A12131.06
5 g, Each
Quantity
Catalog number: A12131.06
also known as A12131-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4644-61-5
IUPAC Name
ethyl 4-oxopiperidine-3-carboxylate hydrochloridyl
Molecular Formula
C8H13ClNO3
InChI Key
OTNOTUGMYUCNQB-UHFFFAOYNA-N
SMILES
[Cl].CCOC(=O)C1CNCCC1=O
Appearance (Color)
White to yellow to cream
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text