Thermo Scientific Chemicals

n-Octylbenzene, 99%, Thermo Scientific Chemicals

Catalog number: A12138.36
500 g, Each
Thermo Scientific Chemicals

n-Octylbenzene, 99%, Thermo Scientific Chemicals

Catalog number: A12138.36
500 g, Each
Quantity
Catalog number: A12138.36
also known as A12138-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2189-60-8
IUPAC Name
octylbenzene
Molecular Formula
C14H22
InChI Key
CDKDZKXSXLNROY-UHFFFAOYSA-N
SMILES
CCCCCCCCC1=CC=CC=C1
Appearance (Color)
Clear colorless
Form
Liquid
Refractive Index
1.4825-1.4855 @ 20?C
Assay (GC)
≥98.5%

Description

n-octylbenzene is used as a solvent to study the self-assembly of derivatives of tetrathiafulvalene (TTF) on graphite and their visualization by scanning tunnelling microscopy (STM). It is also involved in the study the effects and substitution of these compounds. It is also used to prepare charge-transfer complexes with fluoranil and 1,3,5-trinitrobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-octylbenzene is used as a solvent to study the self-assembly of derivatives of tetrathiafulvalene (TTF) on graphite and their visualization by scanning tunnelling microscopy (STM). It is also involved in the study the effects and substitution of these compounds. It is also used to prepare charge-transfer complexes with fluoranil and 1,3,5-trinitrobenzene.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text