Thermo Scientific Chemicals

Phenylacetylene, 98+%, Thermo Scientific Chemicals

Catalog number: A12139.36
500 g, Each
Thermo Scientific Chemicals

Phenylacetylene, 98+%, Thermo Scientific Chemicals

Catalog number: A12139.36
500 g, Each
Quantity
Catalog number: A12139.36
also known as A12139-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
536-74-3
IUPAC Name
ethynylbenzene
Molecular Formula
C8H6
InChI Key
UEXCJVNBTNXOEH-UHFFFAOYSA-N
SMILES
C#CC1=CC=CC=C1
Refractive Index
1.5460-1.5500 @ 20?C
Form
Liquid
Assay (GC)
≥98.0%
Appearance (Color)
Clear colorless to yellow
Identification (FTIR)
Conforms

Description

Phenylacetylene is involved in the preparation of styrene by reduction using Lindlar catalyst. It is used to study the mechanism of the palladium-catalyzed phenyl acetylene oxidative carbonylation reaction. It is also used in the polymerization process to prepare polyphenylacetylene namely 1,2,4-triphenylbenzene and 1,3,5-triphenylbenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylacetylene is involved in the preparation of styrene by reduction using Lindlar catalyst. It is used to study the mechanism of the palladium-catalyzed phenyl acetylene oxidative carbonylation reaction. It is also used in the polymerization process to prepare polyphenylacetylene namely 1,2,4-triphenylbenzene and 1,3,5-triphenylbenzene.

Solubility
Immiscible with water.

Notes
Store in cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, halogens, alkali metals and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text