Thermo Scientific Chemicals

Potassium Dihydrogen Phosphate, ≥98%, Thermo Scientific Chemicals

Catalog number: A12142.36
500 g, Each
Thermo Scientific Chemicals

Potassium Dihydrogen Phosphate, ≥98%, Thermo Scientific Chemicals

Catalog number: A12142.36
500 g, Each
Quantity
Catalog number: A12142.36
also known as A12142-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7778-77-0
IUPAC Name
potassium dihydrogen phosphate
Molecular Formula
H2KO4P
InChI Key
GNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES
[K+].OP(O)([O-])=O
Water Content (Karl Fischer Titration)
≤1.0%
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.0% to ≤102.0%
Solution Test
10% in water: clear
Form
Crystals or powder or crystalline powder

Description

Potassium dihydrogen phosphate is used as a food additive, fertilizer (NPK), paint additive, coating additive, adhesive, buffering agent and in optical modulators. It is extensively used in research and analytical laboratories as a buffer in mobile/sample preparations for analysis by high pressure liquid chromatography (HPLC) and also in organic synthesis. MKP, monopotassium phosphate, owing to its non-linear optical properties, is used in optical modulators and for second-harmonic generation (SHG).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium dihydrogen phosphate is used as a food additive, fertilizer (NPK), paint additive, coating additive, adhesive, buffering agent and in optical modulators. It is extensively used in research and analytical laboratories as a buffer in mobile/sample preparations for analysis by high pressure liquid chromatography (HPLC) and also in organic synthesis. MKP, monopotassium phosphate, owing to its non-linear optical properties, is used in optical modulators and for second-harmonic generation (SHG).

Solubility
Soluble in water. Slightly soluble in ethanol.

Notes
Loses H{2}O at 400°, forming metaphosphateHygroscopic.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text