Thermo Scientific™

trans-Indole-3-acrylic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12143.06
5 g, Each
Thermo Scientific™

trans-Indole-3-acrylic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12143.06
5 g, Each
Quantity
Catalog number: A12143.06
also known as A12143-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29953-71-7
Form
Crystals or powder or crystalline powder
Appearance (Color)
Pale cream to cream to brown
Water Content (Karl Fischer Titration)
≤1.0%
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (HPLC)
≥98.0%

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text