Thermo Scientific Chemicals

4-Iodoanisole, 98+%, Thermo Scientific Chemicals

Catalog number: A12144.22
100 g, Each
Thermo Scientific Chemicals

4-Iodoanisole, 98+%, Thermo Scientific Chemicals

Catalog number: A12144.22
100 g, Each
Quantity
Catalog number: A12144.22
also known as A12144-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
696-62-8
IUPAC Name
1-iodo-4-methoxybenzene
Molecular Formula
C7H7IO
InChI Key
SYSZENVIJHPFNL-UHFFFAOYSA-N
SMILES
COC1=CC=C(I)C=C1
Appearance (Color)
White to cream to pale yellow
Melting Point (clear melt)
47.0-54.0?C
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Assay (GC)
≥98.0%

Description

4-Iodoanisole is used in wide range of medicals industrial applications as well as in human and animal nutrition products, pharmaceutical intermediates, polarizing films for Liquid Crystal Display (LCD) chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodoanisole is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents.

Solubility
Soluble in ethanol, ether, chloroform.

Notes
Light sensitive. Store in the dark. Store away from oxidizing agents, light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text