Thermo Scientific Chemicals

4-tert-Butylthiophenol, 97%, Thermo Scientific Chemicals

Catalog number: A12148.03
1 g, Each
Thermo Scientific Chemicals

4-tert-Butylthiophenol, 97%, Thermo Scientific Chemicals

Catalog number: A12148.03
1 g, Each
Quantity
Catalog number: A12148.03
also known as A12148-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2396-68-1
IUPAC Name
4-tert-butylbenzene-1-thiol
Molecular Formula
C10H14S
InChI Key
GNXBFFHXJDZGEK-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC=C(S)C=C1
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥96.0%
Refractive Index
1.5455-1.5505 @ 20?C
Appearance (Color)
Clear colorless to pale yellow

Description

4-tert-butylthiophenol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-butylthiophenol is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text