Thermo Scientific Chemicals

N-(Benzyloxycarbonyloxy)succinimide, 99%, Thermo Scientific Chemicals

Catalog number: A12153.14
25 g, Each
Thermo Scientific Chemicals

N-(Benzyloxycarbonyloxy)succinimide, 99%, Thermo Scientific Chemicals

Catalog number: A12153.14
25 g, Each
Quantity
Catalog number: A12153.14
also known as A12153-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13139-17-8
IUPAC Name
benzyl 2,5-dioxopyrrolidin-1-yl carbonate
Molecular Formula
C12H11NO5
InChI Key
MJSHDCCLFGOEIK-UHFFFAOYSA-N
SMILES
O=C(OCC1=CC=CC=C1)ON1C(=O)CCC1=O
Assay (HPLC)
≥98.5%
Melting Point (clear melt)
77.0-83.0?C
Appearance (Color)
White
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder

Description

N-(N-Benzyloxycarbonyloxy)succinimide is a reagent used in the preparation of peptide thioacids view peptide precursors. Also used in the preparation of amino glycoside derivatives for the function of topoisomerase for both human and bacterial inhibition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(N-Benzyloxycarbonyloxy)succinimide is a reagent used in the preparation of peptide thioacids view peptide precursors. Also used in the preparation of amino glycoside derivatives for the function of topoisomerase for both human and bacterial inhibition.

Solubility
Soluble in chloroform, methanol, acetone (0.1 g/mL).

Notes
Moisture sensitive. Storage temperature 2-8°C. Store away from oxidizing agents, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text